Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubash3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177823.4 | NP_808491.2 | 5 - 6 | 772 | 102 | |
GenBank mRNA | AK040572.1 | - | 5 - 6 | 733 | 102 | |
AK041690.1 | - | 5 - 6 | 739 | 102 | ||
AK049106.1 | - | 4 - 5 | 672 | 102 | ||
AK169696.1 | - | 5 - 6 | 727 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |