Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175114.3 | NP_780323.2 | 1 - 2 | 235 | 75 | |
GenBank mRNA | BC046285.1 | - | 1 - 2 | 225 | 75 | |
BC058971.1 | - | 1 - 2 | 177 | 6832507 | 75 | |
BY018437.1 | - | 1 - 2 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |