Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyzl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026915.2 | NP_081191.1 | 1 - 2 | 70 | 99 | |
GenBank mRNA | BC024932.1 | - | 1 - 2 | 70 | 3985345 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |