Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164823.1 | NP_001158295.1 | 2 - 3 | 390 | 76 | |
GenBank mRNA | AA939506.1 | - | 1 - 2 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons |