Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xylb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033209.3 | NP_001028381.1 | 4 - 5 | 564 | 84 | |
NM_001199568.1 | NP_001186497.1 | 3 - 4 | 494 | 84 | ||
GenBank mRNA | AK143476.1 | - | 4 - 5 | 564 | 84 | |
AK164186.1 | - | 3 - 4 | 288 | 84 | ||
AK165440.1 | - | 4 - 5 | 361 | 84 | ||
AK165518.1 | - | 4 - 5 | 361 | 84 | ||
AK165759.1 | - | 3 - 4 | 290 | 84 | ||
AK170197.1 | - | 3 - 4 | 307 | 84 | ||
BC025442.1 | - | 4 - 5 | 314 | 84 | ||
BC080743.1 | - | 4 - 5 | 338 | 84 | ||
BC138244.1 | - | 4 - 5 | 513 | 8861263 | 84 | |
BC138247.1 | - | 4 - 5 | 513 | 8861266 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |