Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900013034.mm.1 |
NM_001164567.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013034.mm.1 |
NM_001164567.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013034.mm.1 |
NM_011700.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013034.mm.1 |
NM_011700.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013034.mm.1 |
TC0900001570.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900013032.mm.1 |
NM_001164567.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013032.mm.1 |
NM_001164567.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013032.mm.1 |
NM_011700.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013032.mm.1 |
NM_011700.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013032.mm.1 |
TC0900001570.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900013037.mm.1 |
NM_001164567.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013037.mm.1 |
NM_001164567.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013037.mm.1 |
NM_011700.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013037.mm.1 |
NM_011700.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013037.mm.1 |
TC0900001570.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900013038.mm.1 |
NM_001164567.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013038.mm.1 |
NM_001164567.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013038.mm.1 |
NM_011700.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013038.mm.1 |
NM_011700.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013038.mm.1 |
TC0900001570.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900013030.mm.1 |
NM_001164567.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013030.mm.1 |
NM_001164567.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013030.mm.1 |
NM_011700.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013030.mm.1 |
NM_011700.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013030.mm.1 |
TC0900001570.mm.1 |
MTA TC |