Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vill | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164567.1 | NP_001158039.1 | 6 - 7 | 627 | 62 | |
NM_011700.2 | NP_035830.2 | 6 - 7 | 627 | 62 | ||
GenBank mRNA | AJ344341.1 | - | 6 - 7 | 615 | 62 | |
AK166823.1 | - | 5 - 6 | 859 | 62 | ||
BC021808.1 | - | 6 - 7 | 603 | 3991439 | 62 | |
CF171710.1 | - | 2 - 3 | 184 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |