Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfbr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029575.3 | NP_083851.3 | 1 - 2 | 416 | 97 | |
GenBank mRNA | AF406755.1 | - | 1 - 2 | 90 | 97 | |
AK148167.1 | - | 1 - 2 | 416 | 97 | ||
S69114.1 | - | 1 - 2 | 127 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mus musculus Probe spans exons |