Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400032095.mm.1 |
TC0400003880.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032095.mm.1 |
NM_024412.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032095.mm.1 |
NM_024412.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032090.mm.1 |
TC0400003880.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032090.mm.1 |
NM_001146307.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032090.mm.1 |
NM_001146307.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032090.mm.1 |
NM_024412.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032090.mm.1 |
NM_024412.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032091.mm.1 |
TC0400003880.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032091.mm.1 |
NM_001146307.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032091.mm.1 |
NM_001146307.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032091.mm.1 |
NM_024412.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032091.mm.1 |
NM_024412.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032089.mm.1 |
TC0400003880.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032089.mm.1 |
NM_001146307.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032089.mm.1 |
NM_001146307.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032089.mm.1 |
NM_024412.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032089.mm.1 |
NM_024412.3 |
Refseq |