Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039200.2 | NP_001034289.1 | 9 - 10 | 1055 | 64 | |
NM_146157.4 | NP_666269.2 | 9 - 10 | 1055 | 64 | ||
GenBank mRNA | AK049061.1 | - | 9 - 10 | 1046 | 64 | |
AK077496.1 | - | 9 - 10 | 1046 | 64 | ||
AK089289.1 | - | 9 - 10 | 1034 | 64 | ||
AK129052.1 | - | 9 - 10 | 1008 | 64 | ||
AK161677.1 | - | 9 - 10 | 1018 | 64 | ||
AK170739.1 | - | 9 - 10 | 1034 | 64 | ||
BC048948.1 | - | 8 - 9 | 64 | |||
BC056952.1 | - | 4 - 5 | 503 | 64 | ||
BC129804.1 | - | 9 - 10 | 1042 | 40087024 | 64 | |
BC139366.1 | - | 9 - 10 | 1041 | 8862388 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |