Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024954.1 | NP_001020125.1 | 1 - 2 | 148 | 82 | |
GenBank mRNA | AJ300183.1 | - | 1 - 2 | 76 | 82 | |
AJ300184.1 | - | 1 - 2 | 148 | 82 | ||
AK010506.1 | - | 1 - 2 | 193 | 82 | ||
AK038786.1 | - | 182 | 82 | |||
AK076968.1 | - | 1 - 2 | 203 | 82 | ||
BC032875.1 | - | 1 - 2 | 157 | 1263363 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |