Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024954.1 | NP_001020125.1 | 4 - 5 | 653 | 60 | |
GenBank mRNA | AJ300183.1 | - | 4 - 5 | 581 | 60 | |
BC032875.1 | - | 3 - 4 | 349 | 1263363 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |