Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cope | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021538.1 | NP_067513.1 | 6 - 7 | 604 | 75 | |
GenBank mRNA | AB039837.1 | - | 6 - 7 | 604 | 75 | |
AK003458.1 | - | 6 - 7 | 621 | 75 | ||
AK140753.1 | - | 4 - 5 | 2325 | 75 | ||
BC009170.1 | - | 6 - 7 | 612 | 2812643 | 75 | |
BC057328.1 | - | 4 - 5 | 2290 | 75 | ||
BC083336.1 | - | 6 - 7 | 586 | 5709960 | 75 | |
U89427.1 | - | 2 - 3 | 121 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |