Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134060.4 | NP_598821.1 | 1 - 2 | 110 | 119 | |
GenBank mRNA | AK167051.1 | - | 1 - 2 | 106 | 119 | |
CX238927.1 | - | 1 - 2 | 110 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mus musculus Probe spans exons |