Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpp40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145938.4 | NP_666050.3 | 4 - 5 | 456 | 160 | |
GenBank mRNA | AK048810.1 | - | 456 | 160 | ||
BC024607.1 | - | 4 - 5 | 401 | 4236826 | 160 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |