Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008760.4 | NP_032786.1 | 1 - 2 | 57 | 69 | |
GenBank mRNA | AK028300.1 | - | 1 - 2 | 83 | 69 | |
AK053650.1 | - | 1 - 2 | 58 | 69 | ||
AK160760.1 | - | 1 - 2 | 79 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus 1700 Probe spans exons |