Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175401.3 | NP_780610.2 | 1 - 2 | 105 | 91 | |
GenBank mRNA | AK135553.1 | - | 1 - 2 | 119 | 91 | |
BC040428.1 | - | 1 - 2 | 105 | 3327345 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |