Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK050097.1 | - | 1 - 2 | 115 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |