Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300013262.mm.1 |
NM_001166463.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013262.mm.1 |
NM_001166463.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013262.mm.1 |
NM_001166464.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013262.mm.1 |
NM_001166464.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013262.mm.1 |
NM_001166465.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013262.mm.1 |
NM_001166465.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013262.mm.1 |
NM_001166466.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013262.mm.1 |
NM_001166466.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013262.mm.1 |
NM_009262.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013262.mm.1 |
NM_009262.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013262.mm.1 |
TC1300002086.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300013261.mm.1 |
NM_001166463.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013261.mm.1 |
NM_001166463.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013261.mm.1 |
NM_001166464.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013261.mm.1 |
NM_001166464.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013261.mm.1 |
NM_001166465.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013261.mm.1 |
NM_001166465.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013261.mm.1 |
NM_001166466.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013261.mm.1 |
NM_001166466.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013261.mm.1 |
NM_009262.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013261.mm.1 |
NM_009262.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013261.mm.1 |
TC1300002086.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300013253.mm.1 |
NM_001166463.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013253.mm.1 |
NM_001166463.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013253.mm.1 |
NM_001166464.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013253.mm.1 |
NM_001166464.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013253.mm.1 |
NM_001166465.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013253.mm.1 |
NM_001166465.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013253.mm.1 |
NM_001166466.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013253.mm.1 |
NM_001166466.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013253.mm.1 |
NM_009262.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013253.mm.1 |
NM_009262.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013253.mm.1 |
TC1300002086.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300013260.mm.1 |
NM_001166463.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013260.mm.1 |
NM_001166463.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013260.mm.1 |
NM_001166464.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013260.mm.1 |
NM_001166464.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013260.mm.1 |
NM_001166465.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013260.mm.1 |
NM_001166465.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013260.mm.1 |
NM_001166466.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013260.mm.1 |
NM_001166466.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013260.mm.1 |
NM_009262.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013260.mm.1 |
NM_009262.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013260.mm.1 |
TC1300002086.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300013263.mm.1 |
NM_001166463.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013263.mm.1 |
NM_001166463.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013263.mm.1 |
NM_001166464.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013263.mm.1 |
NM_001166464.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013263.mm.1 |
NM_001166465.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013263.mm.1 |
NM_001166465.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013263.mm.1 |
NM_001166466.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013263.mm.1 |
NM_001166466.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013263.mm.1 |
NM_009262.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013263.mm.1 |
NM_009262.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013263.mm.1 |
TC1300002086.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300013252.mm.1 |
NM_001166463.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013252.mm.1 |
NM_001166463.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013252.mm.1 |
NM_001166464.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013252.mm.1 |
NM_001166464.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013252.mm.1 |
NM_001166465.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013252.mm.1 |
NM_001166465.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013252.mm.1 |
NM_001166466.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013252.mm.1 |
NM_001166466.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013252.mm.1 |
NM_009262.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013252.mm.1 |
NM_009262.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013252.mm.1 |
TC1300002086.mm.1 |
MTA TC |