Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ecm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012324.2 | NP_001012324.1 | 8 - 9 | 1736 | 86 | |
GenBank mRNA | AK038812.1 | - | 8 - 9 | 1598 | 86 | |
AK040995.1 | - | 8 - 9 | 1760 | 86 | ||
AK134746.1 | - | 8 - 9 | 1690 | 86 | ||
AK163501.1 | - | 7 - 8 | 3758 | 86 | ||
BC038144.1 | - | 1 - 2 | 59 | 86 | ||
BC065151.1 | - | 8 - 9 | 1606 | 5708030 | 86 | |
BC089559.1 | - | 8 - 9 | 1736 | 30295705 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |