Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spock3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252620.1 | NP_001239549.1 | 2 - 3 | 255 | 127 | |
NM_023689.3 | NP_076178.1 | 2 - 3 | 369 | 127 | ||
GenBank mRNA | AJ278998.1 | - | 2 - 3 | 263 | 127 | |
AK013644.1 | - | 2 - 3 | 298 | 127 | ||
AK164338.1 | - | 2 - 3 | 243 | 127 | ||
AK220280.1 | - | 2 - 3 | 353 | 127 | ||
BC017601.1 | - | 2 - 3 | 268 | 4502169 | 127 | |
BC053334.1 | - | 2 - 3 | 240 | 6492113 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |