Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fastkd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027123.4 | NP_081399.3 | 3 - 4 | 1654 | 79 | |
GenBank mRNA | AK009264.1 | - | 3 - 4 | 413 | 79 | |
AK031142.1 | - | 3 - 4 | 1641 | 79 | ||
AK032530.1 | - | 3 - 4 | 1640 | 79 | ||
AK033189.1 | - | 3 - 4 | 1655 | 79 | ||
AK036778.1 | - | 3 - 4 | 1640 | 79 | ||
BC042506.1 | - | 3 - 4 | 1631 | 4023412 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |