Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsun2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145354.5 | NP_663329.3 | 14 - 15 | 1674 | 149 | |
GenBank mRNA | AK030124.1 | - | 13 - 14 | 1559 | 149 | |
AK075999.1 | - | 4 - 5 | 453 | 149 | ||
AK150631.1 | - | 14 - 15 | 1628 | 149 | ||
AK151917.1 | - | 14 - 15 | 1628 | 149 | ||
AK154789.1 | - | 14 - 15 | 1666 | 149 | ||
AK171399.1 | - | 14 - 15 | 1631 | 149 | ||
BC013625.1 | - | 10 - 11 | 1146 | 4011674 | 149 | |
BC023735.1 | - | 8 - 9 | 943 | 149 | ||
DQ490066.1 | - | 14 - 15 | 1637 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |