Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134071.3 | NP_598832.3 | 10 - 11 | 1319 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |