Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134071.3 | NP_598832.3 | 13 - 14 | 1774 | 134 | |
GenBank mRNA | BC024900.1 | - | 1 - 2 | 59 | 3152525 | 134 |
BC083095.1 | - | 3 - 4 | 386 | 30458612 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |