Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134071.3 | NP_598832.3 | 14 - 15 | 1929 | 134 | |
GenBank mRNA | BC024900.1 | - | 2 - 3 | 214 | 3152525 | 134 |
BC083095.1 | - | 4 - 5 | 541 | 30458612 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |