Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134071.3 | NP_598832.3 | 5 - 6 | 704 | 125 | |
GenBank mRNA | AK012253.1 | - | 5 - 6 | 697 | 125 | |
AK047775.1 | - | 5 - 6 | 1261 | 125 | ||
BC061188.1 | - | 5 - 6 | 684 | 125 | ||
BC100362.1 | - | 5 - 6 | 676 | 30714028 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |