Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300004496.mm.1 |
NM_001252487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004496.mm.1 |
NM_001252487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004496.mm.1 |
NM_001252488.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004496.mm.1 |
NM_001252488.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004496.mm.1 |
NM_029772.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004496.mm.1 |
NM_029772.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004496.mm.1 |
TC1300000677.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300004501.mm.1 |
NM_001252487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004501.mm.1 |
NM_001252487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004501.mm.1 |
NM_001252488.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004501.mm.1 |
NM_001252488.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004501.mm.1 |
NM_029772.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004501.mm.1 |
NM_029772.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004501.mm.1 |
TC1300000677.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300004495.mm.1 |
NM_001252487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004495.mm.1 |
NM_001252487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004495.mm.1 |
NM_001252488.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004495.mm.1 |
NM_001252488.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004495.mm.1 |
NM_029772.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004495.mm.1 |
NM_029772.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004495.mm.1 |
TC1300000677.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300004498.mm.1 |
NM_001252487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004498.mm.1 |
NM_001252487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004498.mm.1 |
NM_001252488.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004498.mm.1 |
NM_001252488.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004498.mm.1 |
NM_029772.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004498.mm.1 |
NM_029772.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004498.mm.1 |
TC1300000677.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300004500.mm.1 |
NM_001252487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004500.mm.1 |
NM_001252487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004500.mm.1 |
NM_029772.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004500.mm.1 |
NM_029772.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004500.mm.1 |
TC1300000677.mm.1 |
MTA TC |