Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idnk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048060.2 | NP_001041525.1 | 1 - 2 | 92 | 101 | |
NM_198004.3 | NP_932121.2 | 1 - 2 | 92 | 101 | ||
GenBank mRNA | AK017224.1 | - | 1 - 2 | 87 | 101 | |
AK032811.1 | - | 1 - 2 | 75 | 101 | ||
AK040588.1 | - | 1 - 2 | 93 | 101 | ||
AK045763.1 | - | 1 - 2 | 88 | 101 | ||
BC026742.1 | - | 1 - 2 | 61 | 4192995 | 101 | |
BU525297.1 | - | 1 - 2 | 95 | 101 | ||
CX206109.1 | - | 1 - 2 | 68 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |