Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900027042.mm.1 |
NM_001172122.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027042.mm.1 |
NM_001172122.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027042.mm.1 |
NM_001172123.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027042.mm.1 |
NM_001172123.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027042.mm.1 |
NM_001172124.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027042.mm.1 |
NM_001172124.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027042.mm.1 |
NM_001172126.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027042.mm.1 |
NM_001172126.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027042.mm.1 |
NM_178660.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027042.mm.1 |
NM_178660.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027042.mm.1 |
TC0900003204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900027045.mm.1 |
NM_001172122.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027045.mm.1 |
NM_001172122.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027045.mm.1 |
NM_001172123.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027045.mm.1 |
NM_001172123.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027045.mm.1 |
NM_001172124.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027045.mm.1 |
NM_001172124.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027045.mm.1 |
NM_001172126.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027045.mm.1 |
NM_001172126.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027045.mm.1 |
NM_178660.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027045.mm.1 |
NM_178660.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027045.mm.1 |
TC0900003204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900027041.mm.1 |
NM_001172122.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027041.mm.1 |
NM_001172122.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027041.mm.1 |
NM_001172123.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027041.mm.1 |
NM_001172123.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027041.mm.1 |
NM_001172124.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027041.mm.1 |
NM_001172124.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027041.mm.1 |
NM_001172126.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027041.mm.1 |
NM_001172126.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027041.mm.1 |
NM_178660.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027041.mm.1 |
NM_178660.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027041.mm.1 |
TC0900003204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900027043.mm.1 |
NM_001172122.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027043.mm.1 |
NM_001172122.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027043.mm.1 |
NM_001172123.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027043.mm.1 |
NM_001172123.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027043.mm.1 |
NM_001172124.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027043.mm.1 |
NM_001172124.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027043.mm.1 |
NM_001172126.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027043.mm.1 |
NM_001172126.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027043.mm.1 |
NM_178660.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027043.mm.1 |
NM_178660.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027043.mm.1 |
TC0900003204.mm.1 |
MTA TC |