Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbms3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172121.1 | NP_001165592.1 | 5 - 6 | 594 | 82 | |
NM_001172122.1 | NP_001165593.1 | 4 - 5 | 1066 | 82 | ||
NM_001172123.1 | NP_001165594.1 | 4 - 5 | 1066 | 82 | ||
NM_001172124.1 | NP_001165595.1 | 4 - 5 | 1066 | 82 | ||
NM_001172126.1 | NP_001165597.1 | 4 - 5 | 1066 | 82 | ||
NM_178660.4 | NP_848775.2 | 4 - 5 | 1066 | 82 | ||
GenBank mRNA | AK030478.1 | - | 4 - 5 | 726 | 82 | |
AK030609.1 | - | 4 - 5 | 700 | 82 | ||
AK032938.1 | - | 4 - 5 | 1063 | 82 | ||
AK033005.1 | - | 4 - 5 | 709 | 82 | ||
AK041098.1 | - | 4 - 5 | 1068 | 82 | ||
AK046197.1 | - | 5 - 6 | 595 | 82 | ||
AK052196.1 | - | 4 - 5 | 1047 | 82 | ||
BC108402.1 | - | 4 - 5 | 1040 | 6516420 | 82 | |
BC117841.1 | - | 4 - 5 | 506 | 40093225 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |