Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028384.1 | NP_082660.1 | 8 - 9 | 1045 | 103 | |
GenBank mRNA | BM949737.1 | - | 1 - 2 | 133 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |