Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa5b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029401.1 | NP_083677.1 | 2 - 3 | 248 | 80 | |
GenBank mRNA | AK016425.1 | - | 2 - 3 | 269 | 80 | |
BC057376.1 | - | 1 - 2 | 120 | 80 | ||
BC132477.1 | - | 2 - 3 | 248 | 40130754 | 80 | |
BC137851.1 | - | 2 - 3 | 248 | 8860870 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |