Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glp1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021332.2 | NP_067307.2 | 11 - 12 | 1193 | 82 | |
GenBank mRNA | AJ001692.1 | - | 11 - 12 | 1183 | 82 | |
AY293742.1 | - | 11 - 12 | 1183 | 82 | ||
BC103624.1 | - | 11 - 12 | 1182 | 82 | ||
BC105649.1 | - | 11 - 12 | 1182 | 82 | ||
BC105650.1 | - | 11 - 12 | 1182 | 82 | ||
BC139464.1 | - | 11 - 12 | 1220 | 8862486 | 82 | |
BC139466.1 | - | 11 - 12 | 1220 | 8862488 | 82 | |
BC145626.1 | - | 11 - 12 | 1220 | 9054194 | 82 | |
DQ093397.1 | - | 11 - 12 | 1193 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |