Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc74a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166164.1 | NP_001159636.1 | 6 - 7 | 851 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |