Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Susd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163288.2 | NP_001156760.2 | 5 - 6 | 1026 | 83 | |
GenBank mRNA | BC157977.1 | - | 5 - 6 | 951 | 9088138 | 83 |
BC158124.1 | - | 5 - 6 | 1003 | 9007393 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |