Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011673.3 | NP_035803.1 | 2 - 3 | 329 | 149 | |
GenBank mRNA | AK138848.1 | - | 2 - 3 | 345 | 149 | |
BC050828.1 | - | 2 - 3 | 329 | 6512920 | 149 | |
D89866.1 | - | 2 - 3 | 420 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |