Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243192.1 | NP_001230121.1 | 1 - 2 | 124 | 69 | |
GenBank mRNA | AK015611.1 | - | 1 - 2 | 102 | 69 | |
CF732441.1 | - | 1 - 2 | 78 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |