Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019486.1 | NP_062359.1 | 1 | 49 | 91 | |
GenBank mRNA | BC100681.1 | - | 1 | 48 | 91 | |
BC100683.1 | - | 1 | 47 | 91 | ||
BC100684.1 | - | 1 | 48 | 91 | ||
BC100686.1 | - | 1 | 48 | 91 | ||
BC115841.1 | - | 1 | 48 | 91 | ||
BC141882.1 | - | 1 | 49 | 40041946 | 91 | |
BC141892.1 | - | 1 | 49 | 40041948 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |