Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085508.2 | NP_001078977.1 | 2 - 3 | 389 | 95 | |
GenBank mRNA | AK029713.1 | - | 2 - 3 | 390 | 95 | |
BC147569.1 | - | 2 - 3 | 260 | 9056559 | 95 | |
BC147574.1 | - | 2 - 3 | 260 | 9056564 | 95 | |
BC151067.1 | - | 2 - 3 | 260 | 9087980 | 95 | |
BC151070.1 | - | 2 - 3 | 260 | 9087983 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |