Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085508.2 | NP_001078977.1 | 7 - 8 | 1421 | 87 | |
GenBank mRNA | AK029713.1 | - | 7 - 8 | 1422 | 87 | |
BC147569.1 | - | 7 - 8 | 1292 | 9056559 | 87 | |
BC147574.1 | - | 7 - 8 | 1292 | 9056564 | 87 | |
BC151067.1 | - | 6 - 7 | 1023 | 9087980 | 87 | |
BC151070.1 | - | 7 - 8 | 1292 | 9087983 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |