Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163233.1 | NP_001156705.1 | 6 - 7 | 994 | 85 | |
NM_053081.2 | NP_444311.1 | 6 - 7 | 994 | 85 | ||
GenBank mRNA | AF112439.1 | - | 6 - 7 | 803 | 85 | |
AY049715.1 | - | 6 - 7 | 794 | 85 | ||
BC050890.1 | - | 6 - 7 | 1251 | 85 | ||
BC139763.1 | - | 6 - 7 | 797 | 40110334 | 85 | |
BC141282.1 | - | 6 - 7 | 810 | 9055894 | 85 | |
BE292126.1 | - | 1 - 2 | 98 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |