Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183392.2 | NP_899248.1 | 6 - 7 | 1041 | 79 | |
GenBank mRNA | AK087827.1 | - | 6 - 7 | 935 | 79 | |
AK088855.1 | - | 6 - 7 | 966 | 79 | ||
AK133841.1 | - | 6 - 7 | 1039 | 79 | ||
AK150182.1 | - | 6 - 7 | 966 | 79 | ||
AK150841.1 | - | 6 - 7 | 927 | 79 | ||
AK151896.1 | - | 6 - 7 | 927 | 79 | ||
AK152136.1 | - | 6 - 7 | 927 | 79 | ||
AK162037.1 | - | 6 - 7 | 973 | 79 | ||
BC044303.1 | - | 6 - 7 | 969 | 79 | ||
BC068114.1 | - | 6 - 7 | 926 | 5707888 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |