Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025495.3 | NP_079771.1 | 6 - 7 | 681 | 80 | |
GenBank mRNA | AK006234.1 | - | 6 - 7 | 718 | 80 | |
AK012863.1 | - | 6 - 7 | 685 | 80 | ||
BC094280.1 | - | 6 - 7 | 734 | 80 | ||
BC147207.1 | - | 6 - 7 | 661 | 9056262 | 80 | |
BC147208.1 | - | 6 - 7 | 661 | 9056260 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |