Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053248.2 | NP_444478.2 | 13 - 14 | 1733 | 77 | |
GenBank mRNA | AF235001.1 | - | 13 - 14 | 1700 | 77 | |
AF380353.1 | - | 13 - 14 | 1655 | 77 | ||
AK046718.1 | - | 13 - 14 | 1734 | 77 | ||
BC137650.1 | - | 13 - 14 | 1668 | 8860669 | 77 | |
BC137651.1 | - | 13 - 14 | 1668 | 8860670 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |