Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026382.2 | NP_080658.2 | 2 - 3 | 302 | 156 | |
GenBank mRNA | AK007431.1 | - | 2 - 3 | 291 | 156 | |
AK012916.1 | - | 2 - 3 | 297 | 156 | ||
AK018315.1 | - | 2 - 3 | 297 | 156 | ||
AK036713.1 | - | 2 - 3 | 302 | 156 | ||
AK172155.1 | - | 2 - 3 | 291 | 156 | ||
BC021443.1 | - | 2 - 3 | 285 | 5038605 | 156 | |
BC037073.1 | - | 2 - 3 | 292 | 156 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |