Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023842.2 | NP_076331.2 | 12 - 13 | 1905 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |