Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023842.2 | NP_076331.2 | 23 - 24 | 5705 | 147 | |
GenBank mRNA | AK009006.1 | - | 1 - 2 | 1148 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |