Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023842.2 | NP_076331.2 | 3 - 4 | 754 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |