Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctdspl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133710.3 | NP_598471.3 | 1 - 2 | 403 | 83 | |
GenBank mRNA | BC094289.1 | - | 1 - 2 | 88 | 4986338 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |